Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trav14d-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U07881.1 | - | 2 | 245 |
Gene Symbol: Trav14n-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147671.1 | - | 2 | 272 | ||
BC147676.1 | - | 2 | 272 | |||
BC147840.1 | - | 2 | 307 |