Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013725.4 | NP_038753.1 | 3, 4 | 304 | ||
GenBank mRNA | AK005147.1 | - | 3, 4 | 249 | ||
AK011207.1 | - | 807 | ||||
AK150738.1 | - | 3, 4 | 279 | |||
CJ044201.1 | - | 3, 4 | 305 | |||
U93864.1 | - | 3, 4 | 234 |