Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026506.2 | NP_080782.1 | 3 | 161 | ||
GenBank mRNA | AK011129.1 | - | 3 | 160 | ||
AK012802.1 | - | 3 | 150 | |||
AK013235.1 | - | 3 | 162 | |||
AK017852.1 | - | 3 | 510 | |||
BC027499.1 | - | 3 | 134 | 1430048 | ||
BC051470.1 | - | 3 | 206 | 1244608 | ||
BC094411.1 | - | 3 | 148 | 1380524 |