Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007079.3 | NP_009010.2 | 4 | 1253 | ||
NM_032611.2 | NP_116000.1 | 5 | 1328 | |||
XM_005250764.2 | XP_005250821.1 | 7 | 1713 | |||
XM_006716500.2 | XP_006716563.1 | 6 | 1348 | |||
XM_011516803.1 | XP_011515105.1 | 6 | 1590 | |||
XM_011516805.1 | XP_011515107.1 | 7 | 847 | |||
XM_017012998.1 | XP_016868487.1 | 5 | 1345 | |||
XM_017012999.1 | XP_016868488.1 | 5 | 1273 | |||
GenBank mRNA | AF041434.1 | - | 5 | 712 | ||
AJ276554.1 | - | 3 | 440 | |||
BC003105.1 | - | 4 | 734 | 3357244 | ||
BE778111.1 | - | 196 | ||||
BQ637038.1 | - | 4 | 296 | |||
BT007303.1 | - | 4 | 400 |