Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006703.3 | NP_006694.1 | 5 | 677 | ||
GenBank mRNA | AF062529.1 | - | 5 | 512 | ||
AF062530.1 | - | 5 | 512 | |||
AK313440.1 | - | 5 | 580 | |||
BC007727.2 | - | 5 | 638 | 4303483 | ||
BT019984.1 | - | 5 | 359 | |||
BT019985.1 | - | 5 | 359 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 9 | 810 | ||
GenBank mRNA | BU521288.1 | - | 9 | 733 |