Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053982.1 | NP_446434.1 | 4 | 385 | ||
XM_006230097.1 | XP_006230159.1 | 5 | 432 | |||
XM_006230098.1 | XP_006230160.1 | 5 | 475 | |||
GenBank mRNA | BC058452.1 | - | 5 | 410 | 6921126 | |
FQ210209.1 | - | 5 | 407 | |||
FQ211215.1 | - | 5 | 410 | |||
FQ212042.1 | - | 5 | 406 | |||
FQ212257.1 | - | 5 | 404 | |||
FQ214481.1 | - | 5 | 406 | |||
FQ220418.1 | - | 3 | 230 | |||
FQ220927.1 | - | 5 | 402 | |||
FQ220997.1 | - | 5 | 406 | |||
FQ221041.1 | - | 5 | 406 | |||
FQ221112.1 | - | 5 | 414 | |||
FQ221144.1 | - | 5 | 437 | |||
FQ221230.1 | - | 5 | 479 | |||
FQ221308.1 | - | 5 | 408 | |||
FQ221419.1 | - | 5 | 406 | |||
FQ221518.1 | - | 5 | 406 | |||
FQ221677.1 | - | 5 | 404 | |||
FQ221720.1 | - | 5 | 475 | |||
FQ221825.1 | - | 5 | 406 | |||
FQ221840.1 | - | 5 | 406 | |||
FQ221877.1 | - | 5 | 406 | |||
FQ222252.1 | - | 5 | 417 | |||
FQ222272.1 | - | 5 | 406 | |||
FQ222274.1 | - | 5 | 406 | |||
FQ222304.1 | - | 5 | 406 | |||
FQ222376.1 | - | 5 | 406 | |||
FQ222415.1 | - | 5 | 404 | |||
FQ222418.1 | - | 5 | 408 | |||
FQ222718.1 | - | 5 | 406 | |||
FQ222721.1 | - | 5 | 404 | |||
FQ222768.1 | - | 5 | 404 | |||
FQ222892.1 | - | 5 | 407 | |||
FQ222921.1 | - | 5 | 409 | |||
FQ222922.1 | - | 5 | 475 | |||
FQ223006.1 | - | 5 | 406 | |||
FQ223095.1 | - | 5 | 406 | |||
FQ223230.1 | - | 5 | 405 | |||
FQ223341.1 | - | 5 | 406 | |||
FQ223389.1 | - | 5 | 406 | |||
FQ223433.1 | - | 5 | 406 | |||
FQ223787.1 | - | 5 | 406 | |||
FQ223822.1 | - | 5 | 405 | |||
FQ223930.1 | - | 528 | ||||
FQ224201.1 | - | 5 | 406 | |||
FQ224347.1 | - | 5 | 406 | |||
FQ224383.1 | - | 5 | 406 | |||
FQ228419.1 | - | 5 | 405 | |||
FQ228429.1 | - | 5 | 406 | |||
FQ228446.1 | - | 5 | 406 | |||
FQ228475.1 | - | 5 | 407 | |||
FQ228508.1 | - | 5 | 406 | |||
FQ228609.1 | - | 5 | 406 | |||
FQ228617.1 | - | 5 | 405 | |||
FQ229004.1 | - | 5 | 407 | |||
FQ229042.1 | - | 5 | 404 | |||
FQ229063.1 | - | 5 | 405 | |||
X77953.1 | - | 4 | 385 |