Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC681117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760839.1 | XP_008759061.1 | 2 | 362 |
Gene Symbol: Cped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729287.4 | XP_002729333.2 | 15 | 2913 |