Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308045.1 | - | 6 | 735 | ||
BC020721.1 | - | 6 | 735 | 4734153 | ||
K02766.1 | - | 6 | 643 | |||
X02176.1 | - | 6 | 650 |