Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 6 | 1183 | ||
GenBank mRNA | BC099178.1 | - | 6 | 1150 | 7383003 | |
BC099815.1 | - | 6 | 1158 | 7929654 | ||
FQ213300.1 | - | 6 | 1150 | |||
FQ213727.1 | - | 6 | 1143 | |||
FQ214234.1 | - | 4 | 976 | |||
FQ214394.1 | - | 6 | 1142 | |||
FQ214422.1 | - | 6 | 1153 | |||
FQ215102.1 | - | 6 | 1160 | |||
FQ216264.1 | - | 6 | 1176 | |||
FQ216489.1 | - | 6 | 1115 | |||
FQ217912.1 | - | 6 | 1152 | |||
FQ219745.1 | - | 6 | 1218 | |||
FQ219836.1 | - | 6 | 1180 | |||
FQ220601.1 | - | 6 | 1194 | |||
FQ221300.1 | - | 6 | 1124 | |||
FQ223249.1 | - | 6 | 1141 | |||
FQ229179.1 | - | 6 | 1142 | |||
FQ229184.1 | - | 6 | 1116 |
Gene Symbol: LOC100360316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 | 1146 |