Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007085.4 | NP_009016.1 | 9 | 917 | ||
GenBank mRNA | AF288537.1 | - | 3 | 197 | ||
AK289388.1 | - | 9 | 838 | |||
AK291138.1 | - | 9 | 839 | |||
AK300356.1 | - | 8 | 732 | |||
AK300436.1 | - | 9 | 768 | |||
AK312261.1 | - | 9 | 844 | |||
BC000055.2 | - | 9 | 824 | 3505833 | ||
BX647421.1 | - | 9 | 842 | |||
U06863.1 | - | 9 | 833 |