Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053947.1 | NP_446399.1 | 10 | 1150 | ||
XM_008769816.1 | XP_008768038.1 | 11 | 1154 | |||
GenBank mRNA | Z83868.1 | - | 10 | 1150 |