Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007085.4 | NP_009016.1 | 6, 7 | 645 | ||
GenBank mRNA | AK289388.1 | - | 6, 7 | 566 | ||
AK291138.1 | - | 6, 7 | 567 | |||
AK300356.1 | - | 5, 6 | 460 | |||
AK300436.1 | - | 6, 7 | 496 | |||
AK312261.1 | - | 6, 7 | 572 | |||
BC000055.2 | - | 6, 7 | 552 | 3505833 | ||
BX647421.1 | - | 6, 7 | 570 | |||
U06863.1 | - | 6, 7 | 561 |