Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057203.1 | NP_476551.1 | 2 | 145 | ||
GenBank mRNA | AF163477.1 | - | 2 | 90 | ||
AF432871.2 | - | 2 | 122 | |||
BC088218.1 | - | 2 | 145 | 7384271 |