Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 18 | 2429 | ||
NM_007086.3 | NP_009017.1 | 19 | 2522 | |||
XM_006720012.2 | XP_006720075.1 | 19 | 2520 | |||
XM_011536373.2 | XP_011534675.1 | 18 | 2321 | |||
XM_017020940.1 | XP_016876429.1 | 18 | 2433 | |||
XM_017020941.1 | XP_016876430.1 | 16 | 2131 | |||
GenBank mRNA | AJ006266.1 | - | 18 | 2471 | ||
AK001538.1 | - | 7 | 1076 | |||
AK001585.1 | - | 9 | 1445 | |||
AK001620.1 | - | 2 | 249 | |||
AK293005.1 | - | 19 | 2489 | |||
BC063041.1 | - | 19 | 2498 | 6514641 |