Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 24 | 3150 | ||
NM_007086.3 | NP_009017.1 | 25 | 3243 | |||
XM_006720012.2 | XP_006720075.1 | 25 | 3235 | |||
XM_011536373.2 | XP_011534675.1 | 24 | 3042 | |||
XM_017020940.1 | XP_016876429.1 | 24 | 3148 | |||
XM_017020941.1 | XP_016876430.1 | 22 | 2846 | |||
GenBank mRNA | AJ006266.1 | - | 24 | 3192 | ||
AK001538.1 | - | 13 | 1797 | |||
AK001585.1 | - | 15 | 2166 | |||
AK001620.1 | - | 8 | 970 | |||
AK293005.1 | - | 25 | 3210 | |||
BC063041.1 | - | 25 | 3219 | 6514641 |