Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 22 | 2847 | ||
NM_007086.3 | NP_009017.1 | 23 | 2940 | |||
XM_006720012.2 | XP_006720075.1 | 23 | 2932 | |||
XM_011536373.2 | XP_011534675.1 | 22 | 2739 | |||
XM_017020940.1 | XP_016876429.1 | 22 | 2845 | |||
XM_017020941.1 | XP_016876430.1 | 20 | 2543 | |||
GenBank mRNA | AJ006266.1 | - | 22 | 2889 | ||
AK001538.1 | - | 11 | 1494 | |||
AK001585.1 | - | 13 | 1863 | |||
AK001620.1 | - | 6 | 667 | |||
AK293005.1 | - | 23 | 2907 | |||
BC063041.1 | - | 23 | 2916 | 6514641 |