Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007178.3 | NP_009109.3 | 1 | 433 | ||
XM_005253294.1 | XP_005253351.1 | 1 | 431 | |||
GenBank mRNA | AF161496.1 | - | 1 | 380 | ||
AJ010025.1 | - | 1 | 226 | |||
AK297942.1 | - | 1 | 393 | |||
AK312295.1 | - | 1 | 374 | |||
AK315891.1 | - | 1 | 396 | |||
AL136691.1 | - | 1 | 375 | |||
AY049776.1 | - | 1 | 126 | |||
BC000162.2 | - | 1 | 224 | 2900954 | ||
BC062306.1 | - | 1 | 388 | 3349549 | ||
BT020044.1 | - | 1 | 79 | |||
BT020045.1 | - | 1 | 79 | |||
DA666505.1 | - | 1 | 433 | |||
U56430.1 | - | 1 | 132 |