Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144737.2 | NP_653338.2 | 2, 3 | 222 | ||
XM_008769604.1 | XP_008767826.1 | 1, 2 | 486 | |||
XM_008769605.1 | XP_008767827.1 | 2, 3 | 296 | |||
GenBank mRNA | AF458414.1 | - | 2, 3 | 189 | ||
BC070904.1 | - | 2, 3 | 222 | 7100688 | ||
CK365136.1 | - | 2, 3 | 157 |