Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153298.1 | NP_695210.1 | 2 | 175 | ||
XM_008763051.1 | XP_008761273.1 | 2 | 768 | |||
GenBank mRNA | AB056724.1 | - | 2 | 192 |