Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153472.2 | NP_703202.1 | 6, 7 | 739 | ||
GenBank mRNA | AY135567.1 | - | 6, 7 | 694 | ||
BC078712.1 | - | 6, 7 | 739 | 7128354 |