Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173124.1 | NP_775147.1 | 1355 | |||
XM_006241051.2 | XP_006241113.1 | 11 | 1424 | |||
XM_008765157.1 | XP_008763379.1 | 10 | 1372 | |||
XM_008765158.1 | XP_008763380.1 | 11 | 1426 | |||
GenBank mRNA | BC129085.1 | - | 11 | 1316 | 7375658 | |
U39207.1 | - | 1355 |
Gene Symbol: Cyp4f37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271352.1 | NP_001258281.1 | 13 | 1813 |
Gene Symbol: LOC100912391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241095.2 | XP_006241157.1 | 6 | 713 |
Gene Symbol: LOC100912471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241096.2 | XP_006241158.1 | 2249 | |||
XM_006241097.2 | XP_006241159.1 | 9 | 991 |
Gene Symbol: LOC691320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241044.2 | XP_006241106.1 | 6 | 2249 | ||
XM_006241045.1 | XP_006241107.1 | 8 | 958 |