Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013855.1 | NP_001013877.1 | 7, 8 | 955 | ||
XM_006246168.1 | XP_006246230.1 | 8, 9 | 1240 | |||
XM_008767667.1 | XP_008765889.1 | 8, 9 | 1003 | |||
XM_008767668.1 | XP_008765890.1 | 9, 10 | 970 | |||
XM_008767669.1 | XP_008765891.1 | 8, 9 | 1495 | |||
GenBank mRNA | BC089832.1 | - | 7, 8 | 955 | 7324518 |