Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 4 | 985 | ||
XM_005254137.3 | XP_005254194.1 | 4 | 986 | |||
GenBank mRNA | AF140675.1 | - | 4 | 754 | ||
AY327122.1 | - | 4 | 752 |