Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004022.2 | NP_001004022.1 | 7 | 1300 | ||
GenBank mRNA | BC101868.1 | - | 7 | 1300 | 7938504 |