Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 19 | 3700 | ||
XM_005254137.3 | XP_005254194.1 | 19 | 3701 | |||
XM_011521166.2 | XP_011519468.1 | 8 | 1994 | |||
GenBank mRNA | AL110226.1 | - | 4 | 1094 | ||
AY327122.1 | - | 19 | 3467 | |||
BC061631.1 | - | 19 | 3680 | 6650221 |
Gene Symbol: ADAMTS7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045529.3 | 19 | 3216 |