Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175755.2 | NP_786931.1 | 3 | 366 | ||
XM_006248674.1 | XP_006248736.1 | 3 | 471 | |||
XM_008768853.1 | XP_008767075.1 | 3 | 417 | |||
XM_008768854.1 | XP_008767076.1 | 3 | 453 | |||
XM_008768855.1 | XP_008767077.1 | 3 | 471 | |||
GenBank mRNA | AB023634.1 | - | 3 | 366 | ||
BC166854.1 | - | 3 | 471 | 9098491 |