Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000048.1 | NP_001000048.1 | 466 |
Gene Symbol: Olr1562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000047.1 | NP_001000047.1 | 466 |
Gene Symbol: Olr1563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751034.1 | XP_003751082.1 | 1 | 466 |
Gene Symbol: Olr1564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248212.2 | XP_006248274.1 | 1 | 474 |