Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013448.2 | NP_038476.2 | 6, 7 | 1304 | ||
NM_182648.1 | NP_872589.1 | 6, 7 | 1304 | |||
XM_011536374.1 | XP_011534676.1 | 5, 6 | 603 | |||
GenBank mRNA | AB032252.1 | - | 6, 7 | 1301 | ||
AF161435.1 | - | 6, 7 | 990 | |||
AF213467.1 | - | 6, 7 | 902 | |||
AF221130.1 | - | 5, 6 | 736 | |||
BC020636.1 | - | 5, 6 | 1064 | |||
BC108711.1 | - | 6, 7 | 968 |