Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198733.1 | NP_942028.1 | 1, 2 | 184 | ||
GenBank mRNA | AF159245.2 | - | 1, 2 | 184 |