Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011941.1 | NP_001011941.1 | 6 | 854 | ||
XM_006231201.2 | XP_006231263.1 | 7 | 1013 | |||
XM_006231202.2 | XP_006231264.1 | 7 | 1013 | |||
XM_008760322.1 | XP_008758544.1 | 7 | 1013 | |||
GenBank mRNA | BC083761.1 | - | 6 | 854 | 7192975 |