Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013923.1 | NP_001013945.1 | 151 | |||
GenBank mRNA | BC078946.1 | - | 151 | 7109582 |