Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: V1ra14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008898.1 | NP_001008898.1 | 1 | 359 | ||
GenBank mRNA | AY510301.1 | - | 1 | 359 |
Gene Symbol: Vom1r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173298.2 | NP_775420.2 | 1 | 359 | ||
XM_008763052.1 | XP_008761274.1 | 3 | 823 | |||
GenBank mRNA | AY510305.1 | - | 1 | 359 | ||
U36899.1 | - | 3 | 828 |
Gene Symbol: Vom1r103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167553.1 | NP_001161025.1 | 1 | 329 |
Gene Symbol: Vom1r105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173113.1 | NP_775136.1 | 2 | 553 | ||
GenBank mRNA | U36785.1 | - | 2 | 553 |