Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181084.2 | NP_851598.1 | 1 | 220 | ||
XM_006237868.2 | XP_006237930.1 | 2 | 351 | |||
XM_008763528.1 | XP_008761750.1 | 1 | 124 | |||
XM_008763529.1 | XP_008761751.1 | 1 | 124 | |||
GenBank mRNA | AB107917.1 | - | 1 | 220 |