Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765001.1 | XP_008763223.1 | 2 | 2383 | ||
XM_008765002.1 | XP_008763224.1 | 2 | 2306 |