Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106816.1 | NP_001100286.1 | 6 | 993 | ||
XM_006242915.2 | XP_006242977.1 | 6 | 6725 | |||
XM_006242916.2 | XP_006242978.1 | 6 | 1083 |