Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106840.1 | NP_001100310.1 | 213 | |||
GenBank mRNA | BC157819.1 | - | 183 | 5621777 | ||
FJ179399.1 | - | 2, 3 | 146 | |||
FQ230022.1 | - | 193 | ||||
FQ233696.1 | - | 195 | ||||
FQ233840.1 | - | 195 | ||||
X62660.1 | - | 174 |
Gene Symbol: LOC688925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270386.1 | NP_001257315.1 | 1 | 101 | ||
GenBank mRNA | AY387069.1 | - | 1 | 101 |