Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106849.2 | NP_001100319.1 | 3 | 1673 | ||
GenBank mRNA | BC168912.1 | - | 3 | 1673 | 7191741 |