Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106867.1 | NP_001100337.1 | 3 | 405 | ||
GenBank mRNA | BC169125.1 | - | 3 | 325 | 6919533 | |
FQ211778.1 | - | 3 | 408 | |||
FQ214080.1 | - | 3 | 418 |