Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034136.1 | NP_001029308.1 | 3 | 338 | ||
GenBank mRNA | BC100081.1 | - | 3 | 338 | 7325700 | |
FQ214924.1 | - | 3 | 371 | |||
FQ229593.1 | - | 3 | 372 |