Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NISCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276293.1 | NP_001263222.1 | 10, 11 | 1305 | ||
NM_001276294.1 | NP_001263223.1 | 10, 11 | 1305 | |||
NM_007184.3 | NP_009115.2 | 10, 11 | 1305 | |||
GenBank mRNA | AB023192.1 | - | 11, 12 | 1243 | ||
AF082516.1 | - | 10, 11 | 1197 | |||
AK001505.1 | - | 7, 8 | 1418 | |||
AK027428.1 | - | 10, 11 | 1183 | |||
AK291398.1 | - | 10, 11 | 1209 | |||
AK302441.1 | - | 1, 2 | 193 | |||
AK315001.1 | - | 10, 11 | 1209 | |||
BC038102.2 | - | 10, 11 | 1209 | 5268247 | ||
BC054494.1 | - | 10, 11 | 1203 | 6195532 | ||
BC056900.1 | - | 10, 11 | 1175 | 6143316 | ||
BU177208.1 | - | 1, 2 | 56 |