Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 8 | 1445 | ||
NM_001291106.1 | NP_001278035.1 | 8 | 1692 | |||
NM_001291107.1 | NP_001278036.1 | 8 | 1689 | |||
NM_007185.6 | NP_009116.3 | 8 | 1692 | |||
XM_006711139.2 | XP_006711202.1 | 9 | 920 | |||
XM_006711141.2 | XP_006711204.1 | 9 | 917 | |||
XM_011509112.2 | XP_011507414.1 | 7 | 783 | |||
XM_011509114.2 | XP_011507416.1 | 7 | 751 | |||
XM_017000145.1 | XP_016855634.1 | 7 | 758 | |||
XM_017000146.1 | XP_016855635.1 | 7 | 755 | |||
GenBank mRNA | AB209264.1 | - | 8 | 1445 | ||
AF329264.1 | - | 8 | 892 | |||
AK298852.1 | - | 3 | 381 | |||
AY165003.1 | - | 8 | 1682 | |||
BC052491.1 | - | 8 | 1666 | 4820737 | ||
BC068008.1 | - | 7 | 719 | |||
BC104758.1 | - | 8 | 955 | 8143761 | ||
BC143226.1 | - | 8 | 952 | 9051727 | ||
U80746.1 | - | 5 | 571 | |||
U80759.1 | - | 5 | 578 |