Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 1 | 395 | ||
NM_001172649.3 | NP_001166120.1 | 1 | 899 | |||
NM_001291106.1 | NP_001278035.1 | 1 | 899 | |||
NM_001291107.1 | NP_001278036.1 | 1 | 899 | |||
NM_007185.6 | NP_009116.3 | 1 | 899 | |||
XM_006711139.2 | XP_006711202.1 | 2 | 127 | |||
XM_006711141.2 | XP_006711204.1 | 2 | 127 | |||
XM_011509111.2 | XP_011507413.1 | 1 | 747 | |||
GenBank mRNA | AB209264.1 | - | 1 | 395 | ||
AF329264.1 | - | 1 | 99 | |||
AL519587.3 | - | 2 | 131 | |||
AY165003.1 | - | 1 | 889 | |||
BC004145.2 | - | 2 | 77 | |||
BC041088.1 | - | 1 | 131 | |||
BC052491.1 | - | 1 | 873 | 4820737 | ||
BC104758.1 | - | 1 | 162 | 8143761 | ||
BC143226.1 | - | 1 | 162 | 9051727 | ||
DR001924.1 | - | 1 | 493 |