Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191667.1 | NP_001178596.1 | 7 | 1369 | ||
XM_008769234.1 | XP_008767456.1 | 8 | 1578 |