Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107295.1 | NP_001100765.1 | 5 | 566 | ||
XM_006252623.2 | XP_006252685.1 | 5 | 667 | |||
XM_006252624.2 | XP_006252686.1 | 6 | 774 | |||
XM_006252625.2 | XP_006252687.1 | 6 | 760 | |||
XM_006252626.2 | XP_006252688.1 | 5 | 653 | |||
XM_008771000.1 | XP_008769222.1 | 6 | 855 |