Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107326.1 | NP_001100796.1 | 1 | 3473 | ||
XM_006253390.2 | XP_006253452.1 | 2 | 4203 | |||
XM_006253391.2 | XP_006253453.1 | 2 | 4198 | |||
XM_006253392.2 | XP_006253454.1 | 2 | 4041 | |||
XM_006253393.2 | XP_006253455.1 | 2 | 4081 |