Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107475.1 | NP_001100945.1 | 10 | 1692 | ||
XM_006228259.1 | XP_006228321.1 | 9 | 2720 | |||
XM_006228260.1 | XP_006228322.1 | 9 | 2738 | |||
XM_006228261.1 | XP_006228323.1 | 9 | 2721 | |||
XM_006228262.2 | XP_006228324.1 | 9 | 1455 | |||
XM_006228263.1 | XP_006228325.1 | 10 | 2469 | |||
XM_006228264.1 | XP_006228326.1 | 10 | 2452 | |||
XM_006228265.2 | XP_006228327.1 | 9 | 1447 | |||
XM_006228266.1 | XP_006228328.1 | 9 | 2387 | |||
XM_008758923.1 | XP_008757145.1 | 8 | 2989 | |||
XM_008758924.1 | XP_008757146.1 | 9 | 1635 | |||
XM_008758925.1 | XP_008757147.1 | 7 | 2953 | |||
GenBank mRNA | AY724526.1 | - | 8 | 1449 |