Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764909.1 | XP_008763131.1 | 2 | 358 |
Gene Symbol: Dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191740.1 | NP_001178669.1 | 5 | 397 |