Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108051.1 | NP_001101521.1 | 12 | 1460 | ||
XM_006240471.2 | XP_006240533.1 | 15 | 4927 | |||
XM_006240472.2 | XP_006240534.1 | 15 | 4870 | |||
GenBank mRNA | BC168870.1 | - | 14 | 1740 | 7104573 |