Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108089.1 | NP_001101559.1 | 10 | 1624 | ||
GenBank mRNA | BC127512.1 | - | 11 | 1827 | ||
BC161980.1 | - | 10 | 1626 | 9029986 |