Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108198.1 | NP_001101668.1 | 2 | 386 |
Gene Symbol: LOC100909976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773779.1 | XP_008772001.1 | 244 |